Search
Search
| Gene Symbol: SCAI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055761.1 | XP_017911250.1 | 15 - 16 | 1599 | 66 | |
| XM_018055762.1 | XP_017911251.1 | 14 - 15 | 1497 | 66 | ||
| XM_018055763.1 | XP_017911252.1 | 14 - 15 | 1396 | 66 | ||
| GenBank mRNA | JT680672.1 | - | 3 - 4 | 465 | 66 | |
Set Membership: |
|