Search
Search
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059035.1 | XP_017914524.1 | 7 - 8 | 910 | 64 | |
| GenBank mRNA | JO430053.2 | - | 7 - 8 | 721 | 64 | |
| JO632649.2 | - | 7 - 8 | 1064 | 64 | ||
Set Membership: |
|