Search
Search
| Gene Symbol: PPM1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064131.1 | XP_017919620.1 | 4 - 5 | 1118 | 100 | |
| GenBank mRNA | JO394237.2 | - | 4 - 5 | 762 | 100 | |
| JO637989.2 | - | 4 - 5 | 624 | 100 | ||
Set Membership: |
|