Search
Search
| Gene Symbol: DGKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680381.3 | XP_005680438.1 | 22 - 23 | 2531 | 61 | |
| XM_005680383.3 | XP_005680440.1 | 23 - 24 | 2383 | 61 | ||
| XM_018048104.1 | XP_017903593.1 | 23 - 24 | 2271 | 61 | ||
| XM_018048105.1 | XP_017903594.1 | 23 - 24 | 2472 | 61 | ||
| GenBank mRNA | JO612582.2 | - | 1743 | 61 | ||
| JO630152.2 | - | 1 - 2 | 278 | 61 | ||
Set Membership: |
|