Search
Search

TMEM8B| Gene Symbol: TMEM8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052157.1 | XP_017907646.1 | 5 - 6 | 1777 | 65 | |
| XM_018052158.1 | XP_017907647.1 | 5 - 6 | 1777 | 65 | ||
| GenBank mRNA | JO623993.2 | - | 5 - 6 | 1630 | 65 | |
Set Membership: |
|