Search
Search
| Gene Symbol: TBCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065385.1 | XP_017920874.1 | 36 - 37 | 3532 | 68 | |
| XM_018065386.1 | XP_017920875.1 | 35 - 36 | 3443 | 68 | ||
| GenBank mRNA | JO576484.1 | - | 27 - 28 | 2505 | 68 | |
Set Membership: |
|