Search
Search

HSP90AB1| Gene Symbol: HSP90AB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696358.2 | XP_005696415.1 | 6 - 7 | 1101 | 109 | |
| GenBank mRNA | JI861661.1 | - | 5 - 6 | 954 | 109 | |
| JO579192.1 | - | 6 - 7 | 1538 | 109 | ||
| KA344165.1 | - | 6 - 7 | 1064 | 109 | ||
Set Membership: |
|