Search
Search
| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963597.2 | XP_013819051.1 | 10 - 11 | 1548 | 78 | |
| GenBank mRNA | JO499336.2 | - | 2 - 3 | 251 | 78 | |
| JO500738.2 | - | 2 - 3 | 251 | 78 | ||
| JO614942.2 | - | 6 - 7 | 1238 | 78 | ||
Set Membership: |
|