Search
Search

HOMER2| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972981.2 | XP_013828435.1 | 6 - 7 | 972 | 87 | |
| XM_018066348.1 | XP_017921837.1 | 6 - 7 | 846 | 87 | ||
| GenBank mRNA | KA336551.1 | - | 3 - 4 | 1403 | 87 | |
Set Membership: |
|