Search
Search

SMARCC1| Gene Symbol: SMARCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067112.1 | XP_017922601.1 | 17 - 18 | 1826 | 109 | |
| GenBank mRNA | JO581994.1 | - | 17 - 18 | 1898 | 109 | |
Set Membership: |
|