Search
Search

CDKAL1| Gene Symbol: CDKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039026.1 | XP_017894515.1 | 6 - 7 | 599 | 72 | |
| XM_018039027.1 | XP_017894516.1 | 6 - 7 | 600 | 72 | ||
| XM_018039028.1 | XP_017894517.1 | 6 - 7 | 625 | 72 | ||
| XM_018039029.1 | XP_017894518.1 | 5 - 6 | 550 | 72 | ||
| XM_018039030.1 | XP_017894519.1 | 5 - 6 | 555 | 72 | ||
| GenBank mRNA | JO584007.1 | - | 4 - 5 | 459 | 72 | |
Set Membership: |
|