Search
Search
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976202.2 | XP_013831656.1 | 3 - 4 | 396 | 96 | |
| GenBank mRNA | JO583079.1 | - | 1 - 2 | 288 | 96 | |
| KA316751.1 | - | 3 - 4 | 399 | 96 | ||
Set Membership: |
|