Search
Search
| Gene Symbol: MYLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045771.1 | XP_017901260.1 | 12 - 13 | 2203 | 52 | |
| GenBank mRNA | JO397867.2 | - | 3 - 4 | 778 | 52 | |
| JO425463.1 | - | 17 - 18 | 3048 | 52 | ||
| JO425919.1 | - | 17 - 18 | 3048 | 52 | ||
| JO520401.2 | - | 1 - 2 | 82 | 52 | ||
| JO530479.2 | - | 1 - 2 | 82 | 52 | ||
| JO571089.2 | - | 2 - 3 | 367 | 52 | ||
| JO571663.2 | - | 2 - 3 | 367 | 52 | ||
| JO574914.2 | - | 2 - 3 | 367 | 52 | ||
| KA339235.1 | - | 3 - 4 | 778 | 52 | ||
Set Membership: |
|