Search
Search
| Gene Symbol: ASAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055681.1 | XP_017911170.1 | 14 - 15 | 1253 | 62 | |
| XM_018055682.1 | XP_017911171.1 | 13 - 14 | 1244 | 62 | ||
| XM_018055683.1 | XP_017911172.1 | 14 - 15 | 1253 | 62 | ||
| XM_018055685.1 | XP_017911174.1 | 13 - 14 | 1244 | 62 | ||
| XM_018055686.1 | XP_017911175.1 | 14 - 15 | 1253 | 62 | ||
Set Membership: |
|