Search
Search

AKAP10| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064727.1 | XP_017920216.1 | 2 - 3 | 336 | 94 | |
| XM_018064728.1 | XP_017920217.1 | 2 - 3 | 335 | 94 | ||
Set Membership: |
|