Search
Search
| Gene Symbol: FCGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062404.1 | XP_017917893.1 | 18 - 19 | 8359 | 91 | |
| XM_018062405.1 | XP_017917894.1 | 17 - 18 | 7111 | 91 | ||
| GenBank mRNA | KA315085.1 | - | 16 - 17 | 6037 | 91 | |
Set Membership: |
|