Search
Search

C2H2orf76| Gene Symbol: C2H2orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676253.3 | XP_005676310.1 | 3 - 4 | 1114 | 103 | |
| XM_005676254.3 | XP_005676311.1 | 4 - 5 | 426 | 103 | ||
| XM_005676255.3 | XP_005676312.1 | 4 - 5 | 344 | 103 | ||
| GenBank mRNA | JO619775.2 | - | 3 - 4 | 251 | 103 | |
Set Membership: |
|