Search
Search
| Gene Symbol: CAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680210.3 | XP_005680267.2 | 13 - 14 | 3757 | 75 | |
| GenBank mRNA | JO611277.2 | - | 13 - 14 | 3758 | 75 | |
| KA332419.1 | - | 2 - 3 | 661 | 75 | ||
Set Membership: |
|