Search
Search
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692574.3 | XP_005692631.1 | 2 - 3 | 581 | 57 | |
| XM_018062504.1 | XP_017917993.1 | 2 - 3 | 251 | 57 | ||
| GenBank mRNA | JO452910.2 | - | 4 - 5 | 413 | 57 | |
| KA335100.1 | - | 4 - 5 | 467 | 57 | ||
Set Membership: |
|