Search
Search
| Gene Symbol: HEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050126.1 | XP_017905615.1 | 13 - 14 | 3737 | 55 | |
| GenBank mRNA | JO343394.2 | - | 6 - 7 | 1728 | 55 | |
| JO385976.2 | - | 3 - 4 | 272 | 55 | ||
| JO405709.2 | - | 3 - 4 | 268 | 55 | ||
| JO405710.2 | - | 3 - 4 | 268 | 55 | ||
| JO405711.2 | - | 3 - 4 | 268 | 55 | ||
| JO424228.1 | - | 6 - 7 | 5554 | 55 | ||
| JO424229.1 | - | 6 - 7 | 5546 | 55 | ||
| JO424230.1 | - | 6 - 7 | 5554 | 55 | ||
| JO424231.1 | - | 6 - 7 | 5546 | 55 | ||
| JO505348.2 | - | 2 - 3 | 213 | 55 | ||
| JO551144.2 | - | 6 - 7 | 593 | 55 | ||
| JO551146.2 | - | 6 - 7 | 593 | 55 | ||
| JO551149.2 | - | 6 - 7 | 593 | 55 | ||
| JO560036.2 | - | 1 - 2 | 41 | 55 | ||
| JO586536.2 | - | 11 - 12 | 3493 | 55 | ||
| JO586537.2 | - | 11 - 12 | 3505 | 55 | ||
| JO586538.2 | - | 11 - 12 | 3505 | 55 | ||
| JO586539.2 | - | 11 - 12 | 3508 | 55 | ||
| JO586541.2 | - | 11 - 12 | 3505 | 55 | ||
| JO586542.2 | - | 11 - 12 | 3493 | 55 | ||
| JO586543.2 | - | 11 - 12 | 3493 | 55 | ||
| JO586544.2 | - | 11 - 12 | 3508 | 55 | ||
| JO586545.2 | - | 11 - 12 | 3493 | 55 | ||
| JO586546.2 | - | 11 - 12 | 3508 | 55 | ||
| JO586547.2 | - | 11 - 12 | 3505 | 55 | ||
| JO586549.2 | - | 11 - 12 | 3508 | 55 | ||
| JO586550.2 | - | 11 - 12 | 3505 | 55 | ||
| JO586551.2 | - | 11 - 12 | 3508 | 55 | ||
| JO586552.2 | - | 11 - 12 | 3493 | 55 | ||
| KA310875.1 | - | 6 - 7 | 5415 | 55 | ||
Set Membership: |
|