Search
Search
| Gene Symbol: RAB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063867.1 | XP_017919356.1 | 8 - 9 | 809 | 58 | |
| XM_018063868.1 | XP_017919357.1 | 8 - 9 | 928 | 58 | ||
| XM_018063869.1 | XP_017919358.1 | 7 - 8 | 702 | 58 | ||
Set Membership: |
|