Search
Search

CEP112| Gene Symbol: CEP112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063951.1 | XP_017919440.1 | 19 - 20 | 2122 | 59 | |
| XM_018063952.1 | XP_017919441.1 | 19 - 20 | 2142 | 59 | ||
| XM_018063953.1 | XP_017919442.1 | 19 - 20 | 2180 | 59 | ||
| XM_018063954.1 | XP_017919443.1 | 19 - 20 | 2122 | 59 | ||
| XM_018063955.1 | XP_017919444.1 | 19 - 20 | 2124 | 59 | ||
| XM_018063957.1 | XP_017919446.1 | 17 - 18 | 1997 | 59 | ||
| XM_018063958.1 | XP_017919447.1 | 19 - 20 | 2123 | 59 | ||
| XM_018063959.1 | XP_017919448.1 | 19 - 20 | 2121 | 59 | ||
| XM_018063960.1 | XP_017919449.1 | 14 - 15 | 1381 | 59 | ||
| XM_018063961.1 | XP_017919450.1 | 17 - 18 | 1838 | 59 | ||
| XM_018063962.1 | XP_017919451.1 | 12 - 13 | 1583 | 59 | ||
| XM_018063964.1 | XP_017919453.1 | 12 - 13 | 1280 | 59 | ||
| XM_018063965.1 | XP_017919454.1 | 3 - 4 | 356 | 59 | ||
| XM_018063966.1 | XP_017919455.1 | 3 - 4 | 559 | 59 | ||
| XM_018063968.1 | XP_017919457.1 | 2 - 3 | 393 | 59 | ||
| GenBank mRNA | JO639162.2 | - | 2 - 3 | 241 | 59 | |
| JO639163.2 | - | 2 - 3 | 241 | 59 | ||
Set Membership: |
|