Search
Search
| Gene Symbol: PLIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050876.1 | XP_017906365.1 | 6 - 7 | 5023 | 66 | |
| XM_018050877.1 | XP_017906366.1 | 7 - 8 | 4924 | 66 | ||
| GenBank mRNA | JO362555.2 | - | 2 - 3 | 665 | 66 | |
Set Membership: |
|