Search
Search
| Gene Symbol: TMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054201.1 | XP_017909690.1 | 4 - 5 | 606 | 65 | |
| XM_018054202.1 | XP_017909691.1 | 4 - 5 | 839 | 65 | ||
| GenBank mRNA | JO623800.2 | - | 6 - 7 | 1402 | 65 | |
Set Membership: |
|