Search
Search

CCDC92| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061369.1 | XP_017916858.1 | 1 - 2 | 1255 | 57 | |
| XM_018061370.1 | XP_017916859.1 | 2 - 3 | 242 | 57 | ||
| XM_018061371.1 | XP_017916860.1 | 3 - 4 | 316 | 57 | ||
| GenBank mRNA | JT744834.1 | - | 1 - 2 | 265 | 57 | |
Set Membership: |
|