Search
Search
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972587.2 | XP_013828041.1 | 12 - 13 | 2547 | 64 | |
| XM_013972588.2 | XP_013828042.1 | 10 - 11 | 1886 | 64 | ||
| XM_013972589.2 | XP_013828043.1 | 9 - 10 | 1430 | 64 | ||
| XM_018064928.1 | XP_017920417.1 | 11 - 12 | 1949 | 64 | ||
Set Membership: |
|