Search
Search
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972587.2 | XP_013828041.1 | 4 - 5 | 1207 | 82 | |
| XM_013972588.2 | XP_013828042.1 | 3 - 4 | 609 | 82 | ||
| XM_018064928.1 | XP_017920417.1 | 3 - 4 | 609 | 82 | ||
Set Membership: |
|