Search
Search

TBC1D30| Gene Symbol: TBC1D30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963894.2 | XP_013819348.2 | 13 - 14 | 2955 | 84 | |
| XM_018047985.1 | XP_017903474.1 | 12 - 13 | 2256 | 84 | ||
| GenBank mRNA | JO427569.1 | - | 13 - 14 | 1683 | 84 | |
Set Membership: |
|