Search
Search
| Gene Symbol: XPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052206.1 | XP_017907695.1 | 4 - 5 | 624 | 116 | |
| XM_018052208.1 | XP_017907697.1 | 4 - 5 | 624 | 116 | ||
| GenBank mRNA | JO584595.1 | - | 3 - 4 | 480 | 116 | |
| KA340672.1 | - | 2 - 3 | 1104 | 116 | ||
Set Membership: |
|