Search
Search

MAPK8IP3| Gene Symbol: MAPK8IP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040036.1 | XP_017895525.1 | 15 - 16 | 2370 | 67 | |
| XM_018040037.1 | XP_017895526.1 | 15 - 16 | 2367 | 67 | ||
| XM_018040038.1 | XP_017895527.1 | 14 - 15 | 2352 | 67 | ||
| XM_018040039.1 | XP_017895528.1 | 14 - 15 | 2346 | 67 | ||
| XM_018040040.1 | XP_017895529.1 | 14 - 15 | 2343 | 67 | ||
| XM_018040041.1 | XP_017895530.1 | 15 - 16 | 2343 | 67 | ||
| XM_018040042.1 | XP_017895531.1 | 13 - 14 | 2340 | 67 | ||
| XM_018040043.1 | XP_017895532.1 | 13 - 14 | 2325 | 67 | ||
| XM_018040044.1 | XP_017895533.1 | 13 - 14 | 2301 | 67 | ||
| XM_018040045.1 | XP_017895534.1 | 15 - 16 | 2277 | 67 | ||
| XM_018040046.1 | XP_017895535.1 | 15 - 16 | 2274 | 67 | ||
| XM_018040047.1 | XP_017895536.1 | 14 - 15 | 2253 | 67 | ||
| XM_018040048.1 | XP_017895537.1 | 14 - 15 | 2250 | 67 | ||
| XM_018040049.1 | XP_017895538.1 | 13 - 14 | 2235 | 67 | ||
| XM_018040050.1 | XP_017895539.1 | 10 - 11 | 1026 | 67 | ||
Set Membership: |
|