Search
Search
| Gene Symbol: SUMF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040603.1 | XP_017896092.1 | 8 - 9 | 899 | 63 | |
| XM_018040604.1 | XP_017896093.1 | 8 - 9 | 896 | 63 | ||
| GenBank mRNA | JO588165.2 | - | 1 - 2 | 484 | 63 | |
| KA325609.1 | - | 1 - 2 | 582 | 63 | ||
Set Membership: |
|