Search
Search

| Gene Symbol: POLR3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681111.3 | XP_005681168.1 | 5 - 6 | 883 | 63 | |
| GenBank mRNA | JO428297.1 | - | 1 - 2 | 1703 | 63 | |
| JO577759.1 | - | 1 - 2 | 1721 | 63 | ||
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO634090.2 | - | 18 - 19 | 4142 | 63 | |
Set Membership: |
|