Search
Search

ALDH7A1| Gene Symbol: ALDH7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682692.3 | XP_005682749.1 | 13 - 14 | 1369 | 62 | |
| GenBank mRNA | JO576448.1 | - | 12 - 13 | 1249 | 62 | |
Set Membership: |
|