Search
Search
| Gene Symbol: CDK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687557.3 | XP_005687614.1 | 4 - 5 | 1040 | 72 | |
| XM_018056580.1 | XP_017912069.1 | 4 - 5 | 1041 | 72 | ||
| GenBank mRNA | JO515064.2 | - | 4 - 5 | 495 | 72 | |
Set Membership: |
|