Search
Search
| Gene Symbol: PSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699158.3 | XP_005699215.3 | 14 - 15 | 1643 | 66 | |
| XM_018042179.1 | XP_017897668.1 | 14 - 15 | 1640 | 66 | ||
| XM_018042181.1 | XP_017897670.1 | 13 - 14 | 1634 | 66 | ||
| GenBank mRNA | JO425234.1 | - | 7 - 8 | 2408 | 66 | |
Set Membership: |
|