Search
Search
| Gene Symbol: MBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048023.1 | XP_017903512.1 | 4 - 5 | 591 | 74 | |
| XM_018048024.1 | XP_017903513.1 | 4 - 5 | 340 | 74 | ||
| GenBank mRNA | JO447445.2 | - | 4 - 5 | 562 | 74 | |
Set Membership: |
|