Search
Search
| Gene Symbol: MAU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051490.1 | XP_017906979.1 | 13 - 14 | 1348 | 77 | |
| GenBank mRNA | JO595646.2 | - | 12 - 13 | 2629 | 77 | |
| JO644256.2 | - | 6 - 7 | 2416 | 77 | ||
Set Membership: |
|