Search
Search

SLC27A2| Gene Symbol: SLC27A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685790.3 | XP_005685847.2 | 3 - 4 | 1038 | 76 | |
| GenBank mRNA | JO427913.1 | - | 3 - 4 | 1289 | 76 | |
Set Membership: |
|