Search
Search

FASTKD3| Gene Symbol: FASTKD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694867.3 | XP_005694924.1 | 2 - 3 | 1829 | 97 | |
| XM_018065769.1 | XP_017921258.1 | 2 - 3 | 1571 | 97 | ||
Set Membership: |
|