Search
Search

BAIAP2L1| Gene Symbol: BAIAP2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040635.1 | XP_017896124.1 | 12 - 13 | 1680 | 59 | |
| GenBank mRNA | KA349051.1 | - | 12 - 13 | 1638 | 59 | |
Set Membership: |
|