Search
Search
| Gene Symbol: WAPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699311.3 | XP_005699368.1 | 14 - 15 | 3422 | 83 | |
| XM_018042447.1 | XP_017897936.1 | 14 - 15 | 3409 | 83 | ||
| GenBank mRNA | JI861778.1 | - | 12 - 13 | 2806 | 83 | |
Set Membership: |
|