Search
Search

C29H11orf54| Gene Symbol: C29H11orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699392.3 | XP_005699449.2 | 5 - 6 | 542 | 99 | |
| GenBank mRNA | JO430620.2 | - | 4 - 5 | 1557 | 99 | |
Set Membership: |
|