Search
Search

SNRNP200| Gene Symbol: SNRNP200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054846.1 | XP_017910335.1 | 13 - 14 | 1791 | 56 | |
| GenBank mRNA | JO555646.2 | - | 1 - 2 | 226 | 56 | |
Set Membership: |
|