Search
Search
| Gene Symbol: ASB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055950.1 | XP_017911439.1 | 2 - 3 | 430 | 57 | |
| XM_018055951.1 | XP_017911440.1 | 2 - 3 | 430 | 57 | ||
| GenBank mRNA | JO628185.2 | - | 4 - 5 | 1783 | 57 | |
Set Membership: |
|