Search
Search
| Gene Symbol: HK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050630.1 | XP_017906119.1 | 6 - 7 | 1382 | 64 | |
| XM_018050631.1 | XP_017906120.1 | 3 - 4 | 280 | 64 | ||
| GenBank mRNA | JO338923.2 | - | 5 - 6 | 1057 | 64 | |
Set Membership: |
|