Search
Search
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686519.3 | XP_005686576.1 | 24 - 25 | 4328 | 64 | |
| XM_005686520.3 | XP_005686577.1 | 23 - 24 | 4057 | 64 | ||
| XM_005686521.3 | XP_005686578.1 | 23 - 24 | 4272 | 64 | ||
| XM_005686522.3 | XP_005686579.1 | 24 - 25 | 4113 | 64 | ||
| XM_018055146.1 | XP_017910635.1 | 24 - 25 | 4325 | 64 | ||
Set Membership: |
|