Search
Search
| Gene Symbol: PAX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055344.1 | XP_017910833.1 | 11 - 12 | 1466 | 65 | |
| XM_018055345.1 | XP_017910834.1 | 11 - 12 | 1463 | 65 | ||
| XM_018055346.1 | XP_017910835.1 | 10 - 11 | 1259 | 65 | ||
| XM_018055347.1 | XP_017910836.1 | 10 - 11 | 1256 | 65 | ||
| GenBank mRNA | JO427594.1 | - | 1 - 2 | 1203 | 65 | |
Set Membership: |
|