Search
Search
| Gene Symbol: MBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039537.1 | XP_017895026.1 | 3 - 4 | 1076 | 69 | |
| GenBank mRNA | JO579824.1 | - | 4 - 5 | 1085 | 69 | |
| KA343712.1 | - | 4 - 5 | 1139 | 69 | ||
Set Membership: |
|