Search
Search
| Gene Symbol: ABCC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040950.1 | XP_017896439.1 | 31 - 32 | 4643 | 57 | |
| XM_018040951.1 | XP_017896440.1 | 30 - 31 | 4591 | 57 | ||
| GenBank mRNA | JO410065.2 | - | 1 - 2 | 729 | 57 | |
Set Membership: |
|