Search
Search
| Gene Symbol: NCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675077.3 | XP_005675134.1 | 2 - 3 | 345 | 78 | |
| GenBank mRNA | JO324379.2 | - | 2 - 3 | 315 | 78 | |
| JO344986.2 | - | 2 - 3 | 253 | 78 | ||
| JO411267.2 | - | 2 - 3 | 326 | 78 | ||
| JO449525.2 | - | 2 - 3 | 385 | 78 | ||
| JO521828.2 | - | 2 - 3 | 294 | 78 | ||
| JO550510.2 | - | 2 - 3 | 342 | 78 | ||
| JO570212.2 | - | 2 - 3 | 322 | 78 | ||
| JO579684.1 | - | 2 - 3 | 484 | 78 | ||
| JO641339.2 | - | 1 - 2 | 166 | 78 | ||
| JO641340.2 | - | 2 - 3 | 288 | 78 | ||
| JT658581.1 | - | 4 - 5 | 588 | 78 | ||
| JT737794.1 | - | 2 - 3 | 296 | 78 | ||
| KA336667.1 | - | 333 | 78 | |||
Set Membership: |
|